The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033026.1 | NP_001028198.1 | 6 - 7 | 1091 | 55 |
NM_033420.3 | NP_219488.1 | 6 - 7 | 1091 | 55 | |
XM_005259675.2 | XP_005259732.1 | 6 - 7 | 1136 | 55 | |
XM_005259676.2 | XP_005259733.1 | 6 - 7 | 1118 | 55 | |
XM_005259677.2 | XP_005259734.1 | 6 - 7 | 1136 | 55 | |
XM_005259678.2 | XP_005259735.1 | 6 - 7 | 1136 | 55 | |
XM_017027457.1 | XP_016882946.1 | 6 - 7 | 1136 | 55 | |
GenBank mRNA | AK090400.1 | - | 2 - 3 | 156 | 55 |
BC008957.2 | - | 6 - 7 | 1064 | 55 | |
DQ005958.1 | - | 6 - 7 | 1073 | 55 |