Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAF5
Entrez Gene ID
8816
Gene Name
DDB1 and CUL4 associated factor 5
Gene Aliases
BCRG2, BCRP2, D14S1461E, WDR22
Chromosome Location
Chr.14: 69050920 - 69153393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284206.1 NP_001271135.1 4 - 5 748 95
NM_001284207.1 NP_001271136.1 4 - 5 504 95
NM_001284208.1 NP_001271137.1 4 - 5 751 95
NM_003861.2 NP_003852.1 4 - 5 751 95
XM_006720297.2 XP_006720360.1 4 - 5 751 95
XM_006720298.2 XP_006720361.1 4 - 5 748 95
XM_006720299.3 XP_006720362.1 4 - 5 386 95
XM_006720300.2 XP_006720363.1 4 - 5 407 95
XM_011537278.1 XP_011535580.1 5 - 6 1020 95
XM_011537279.2 XP_011535581.1 4 - 5 483 95
XM_017021733.1 XP_016877222.1 4 - 5 341 95
XM_017021734.1 XP_016877223.1 4 - 5 403 95
XM_017021735.1 XP_016877224.1 4 - 5 483 95
XM_017021736.1 XP_016877225.1 4 - 5 407 95
XM_017021738.1 XP_016877227.1 4 - 5 359 95
XM_017021739.1 XP_016877228.1 4 - 5 446 95
GenBank mRNA AB058727.1 - 4 - 5 586 95
AL831932.2 - 4 - 5 504 95
BC022967.1 - 4 - 5 538 95
BC136632.1 - 4 - 5 680 95
BX648006.1 - 4 - 5 555 95

View Full Product Details