Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032291.3 NP_115667.2 14 - 15 1216 70
XM_005271264.3 XP_005271321.1 14 - 15 1050 70
XM_006710961.2 XP_006711024.1 14 - 15 1050 70
XM_006710966.2 XP_006711029.1 14 - 15 1050 70
XM_006710967.2 XP_006711030.1 14 - 15 1038 70
XM_006710969.2 XP_006711032.1 13 - 14 966 70
XM_017002505.1 XP_016857994.1 14 - 15 1038 70
XM_017002506.1 XP_016857995.1 15 - 16 1161 70
XM_017002507.1 XP_016857996.1 14 - 15 1213 70
XM_017002508.1 XP_016857997.1 15 - 16 1159 70
XM_017002509.1 XP_016857998.1 12 - 13 1014 70
XM_017002510.1 XP_016857999.1 13 - 14 1137 70
XM_017002511.1 XP_016858000.1 15 - 16 1161 70
XM_017002512.1 XP_016858001.1 15 - 16 1034 70
XM_017002513.1 XP_016858002.1 14 - 15 1213 70
XM_017002514.1 XP_016858003.1 13 - 14 966 70
XM_017002515.1 XP_016858004.1 11 - 12 942 70
XM_017002516.1 XP_016858005.1 12 - 13 1065 70
XM_017002517.1 XP_016858006.1 15 - 16 1160 70
XM_017002518.1 XP_016858007.1 14 - 15 1088 70
XM_017002519.1 XP_016858008.1 14 - 15 1113 70
XM_017002520.1 XP_016858009.1 12 - 13 954 70
XM_017002521.1 XP_016858010.1 13 - 14 966 70
XM_017002522.1 XP_016858011.1 14 - 15 1088 70
XM_017002523.1 XP_016858012.1 14 - 15 1112 70
GenBank mRNA AB210039.1 - 14 - 15 998 70
AL136561.1 - 14 - 15 890 70
BC040516.1 - 15 - 16 963 70

View Full Product Details