The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005934.3 | NP_005925.2 | 5 - 6 | 716 | 73 |
XM_011528021.1 | XP_011526323.1 | 5 - 6 | 690 | 73 | |
XM_011528022.1 | XP_011526324.1 | 5 - 6 | 573 | 73 | |
XM_017026819.1 | XP_016882308.1 | 5 - 6 | 1272 | 73 | |
GenBank mRNA | D14539.1 | - | 5 - 6 | 716 | 73 |
L04285.1 | - | 4 - 5 | 552 | 73 |