The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001554.4 | NP_001545.2 | 1 - 2 | 287 | 90 |
GenBank mRNA | AF003594.1 | - | 1 - 2 | 186 | 90 |
AF031385.1 | - | 1 - 2 | 143 | 90 | |
AK223385.1 | - | 1 - 2 | 288 | 90 | |
AK314571.1 | - | 1 - 2 | 288 | 90 | |
BC001271.1 | - | 1 - 2 | 222 | 90 | |
BC009199.2 | - | 1 - 2 | 271 | 90 | |
BC016952.1 | - | 1 - 2 | 278 | 90 | |
CR536519.1 | - | 1 - 2 | 63 | 90 | |
DA764894.1 | - | 1 - 2 | 289 | 90 | |
U62015.1 | - | 1 - 2 | 272 | 90 | |
Y11307.1 | - | 1 - 2 | 286 | 90 | |
Y12084.1 | - | 1 - 2 | 277 | 90 | |
Z98053.1 | - | 1 - 2 | 281 | 90 |