The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195573.1 | NP_001182502.1 | 8 - 9 | 1557 | 84 |
NM_001271282.2 | NP_001258211.1 | 9 - 10 | 1745 | 84 | |
NM_001291628.1 | NP_001278557.1 | 9 - 10 | 1633 | 84 | |
NM_030621.4 | NP_085124.2 | 11 - 12 | 1804 | 84 | |
NM_177438.2 | NP_803187.1 | 9 - 10 | 1750 | 84 | |
XM_011536599.1 | XP_011534901.1 | 11 - 12 | 1936 | 84 | |
XM_011536600.2 | XP_011534902.1 | 11 - 12 | 1811 | 84 | |
XM_011536601.2 | XP_011534903.1 | 12 - 13 | 1956 | 84 | |
XM_011536602.2 | XP_011534904.1 | 12 - 13 | 1847 | 84 | |
XM_011536604.2 | XP_011534906.1 | 8 - 9 | 1632 | 84 | |
XM_011536605.2 | XP_011534907.1 | 2 - 3 | 6437 | 84 | |
XM_017021120.1 | XP_016876609.1 | 11 - 12 | 2243 | 84 | |
XM_017021121.1 | XP_016876610.1 | 9 - 10 | 1638 | 84 | |
XM_017021122.1 | XP_016876611.1 | 10 - 11 | 1682 | 84 | |
XM_017021123.1 | XP_016876612.1 | 10 - 11 | 2113 | 84 | |
GenBank mRNA | AB023145.2 | - | 9 - 10 | 1750 | 84 |
AJ132261.2 | - | 9 - 10 | 1753 | 84 | |
BC150287.1 | - | 9 - 10 | 1750 | 84 | |
HM595745.1 | - | 8 - 9 | 1521 | 84 |