The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195573.1 | NP_001182502.1 | 11 - 12 | 2085 | 114 |
NM_001271282.2 | NP_001258211.1 | 12 - 13 | 2273 | 114 | |
NM_001291628.1 | NP_001278557.1 | 12 - 13 | 2161 | 114 | |
NM_030621.4 | NP_085124.2 | 14 - 15 | 2332 | 114 | |
NM_177438.2 | NP_803187.1 | 12 - 13 | 2278 | 114 | |
XM_011536599.1 | XP_011534901.1 | 14 - 15 | 2464 | 114 | |
XM_011536600.2 | XP_011534902.1 | 14 - 15 | 2339 | 114 | |
XM_011536601.2 | XP_011534903.1 | 15 - 16 | 2484 | 114 | |
XM_011536602.2 | XP_011534904.1 | 15 - 16 | 2375 | 114 | |
XM_011536604.2 | XP_011534906.1 | 11 - 12 | 2160 | 114 | |
XM_011536605.2 | XP_011534907.1 | 5 - 6 | 6965 | 114 | |
XM_017021120.1 | XP_016876609.1 | 14 - 15 | 2771 | 114 | |
XM_017021121.1 | XP_016876610.1 | 12 - 13 | 2166 | 114 | |
XM_017021122.1 | XP_016876611.1 | 13 - 14 | 2210 | 114 | |
XM_017021123.1 | XP_016876612.1 | 13 - 14 | 2641 | 114 | |
GenBank mRNA | AB023145.2 | - | 12 - 13 | 2278 | 114 |
AB028449.1 | - | 13 - 14 | 2228 | 114 | |
AJ132261.2 | - | 12 - 13 | 2281 | 114 | |
BC150287.1 | - | 12 - 13 | 2278 | 114 | |
HM595745.1 | - | 11 - 12 | 2049 | 114 |