The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195573.1 | NP_001182502.1 | 7 - 8 | 1423 | 72 |
NM_001271282.2 | NP_001258211.1 | 8 - 9 | 1611 | 72 | |
NM_001291628.1 | NP_001278557.1 | 8 - 9 | 1499 | 72 | |
NM_030621.4 | NP_085124.2 | 10 - 11 | 1670 | 72 | |
NM_177438.2 | NP_803187.1 | 8 - 9 | 1616 | 72 | |
XM_011536599.1 | XP_011534901.1 | 10 - 11 | 1802 | 72 | |
XM_011536600.2 | XP_011534902.1 | 10 - 11 | 1677 | 72 | |
XM_011536601.2 | XP_011534903.1 | 11 - 12 | 1822 | 72 | |
XM_011536602.2 | XP_011534904.1 | 11 - 12 | 1713 | 72 | |
XM_011536604.2 | XP_011534906.1 | 7 - 8 | 1498 | 72 | |
XM_017021120.1 | XP_016876609.1 | 10 - 11 | 2109 | 72 | |
XM_017021121.1 | XP_016876610.1 | 8 - 9 | 1504 | 72 | |
XM_017021122.1 | XP_016876611.1 | 9 - 10 | 1548 | 72 | |
XM_017021123.1 | XP_016876612.1 | 9 - 10 | 1979 | 72 | |
GenBank mRNA | AB023145.2 | - | 8 - 9 | 1616 | 72 |
AB028449.1 | - | 9 - 10 | 1566 | 72 | |
AJ132261.2 | - | 8 - 9 | 1619 | 72 | |
BC150287.1 | - | 8 - 9 | 1616 | 72 | |
HM595745.1 | - | 7 - 8 | 1387 | 72 |