The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006035.3 | NP_006026.3 | 19 - 20 | 2956 | 139 |
XM_005268227.1 | XP_005268284.1 | 19 - 20 | 2786 | 139 | |
XM_005268228.1 | XP_005268285.1 | 19 - 20 | 2786 | 139 | |
XM_005268229.1 | XP_005268286.1 | 19 - 20 | 2786 | 139 | |
XM_005268230.4 | XP_005268287.1 | 19 - 20 | 2786 | 139 | |
XM_011537387.1 | XP_011535689.1 | 19 - 20 | 2786 | 139 | |
GenBank mRNA | AB032950.3 | - | 19 - 20 | 2947 | 139 |
AF128625.1 | - | 19 - 20 | 3013 | 139 | |
BC155541.1 | - | 19 - 20 | 2959 | 139 | |
BX248752.1 | - | 1 - 2 | 137 | 139 |