The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006035.3 | NP_006026.3 | 2 - 3 | 498 | 133 |
XM_005268227.1 | XP_005268284.1 | 2 - 3 | 328 | 133 | |
XM_005268228.1 | XP_005268285.1 | 2 - 3 | 328 | 133 | |
XM_005268229.1 | XP_005268286.1 | 2 - 3 | 328 | 133 | |
XM_005268230.4 | XP_005268287.1 | 2 - 3 | 328 | 133 | |
XM_011537387.1 | XP_011535689.1 | 2 - 3 | 328 | 133 | |
GenBank mRNA | AB032950.3 | - | 2 - 3 | 489 | 133 |
AF128625.1 | - | 2 - 3 | 555 | 133 | |
BC047871.1 | - | 2 - 3 | 391 | 133 | |
BC048261.1 | - | 2 - 3 | 423 | 133 | |
BC155541.1 | - | 2 - 3 | 501 | 133 |