Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC42BPB
Entrez Gene ID
9578
Gene Name
CDC42 binding protein kinase beta
Gene Aliases
MRCKB
Chromosome Location
Chr.14: 102932379 - 103057405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006035.3 NP_006026.3 35 - 36 5166 78
XM_005268227.1 XP_005268284.1 36 - 37 5047 78
XM_005268228.1 XP_005268285.1 35 - 36 4969 78
XM_005268229.1 XP_005268286.1 34 - 35 4918 78
GenBank mRNA AB032950.3 - 35 - 36 5157 78
AF128625.1 - 35 - 36 5223 78
BC155541.1 - 35 - 36 5169 78

View Full Product Details