The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018043.5 | NP_060513.5 | 20 - 21 | 2355 | 69 |
NR_030691.1 | 18 - 19 | 2113 | 69 | ||
XM_006718602.2 | XP_006718665.1 | 21 - 22 | 2137 | 69 | |
XM_006718604.2 | XP_006718667.1 | 21 - 22 | 2252 | 69 | |
XM_006718605.2 | XP_006718668.1 | 18 - 19 | 2108 | 69 | |
XM_011545121.2 | XP_011543423.1 | 21 - 22 | 2129 | 69 | |
XM_011545123.2 | XP_011543425.1 | 21 - 22 | 2126 | 69 | |
XM_011545124.2 | XP_011543426.1 | 20 - 21 | 2117 | 69 | |
XM_011545125.2 | XP_011543427.1 | 20 - 21 | 2051 | 69 | |
XM_011545126.2 | XP_011543428.1 | 19 - 20 | 2039 | 69 | |
XM_011545127.2 | XP_011543429.1 | 19 - 20 | 1985 | 69 | |
XM_011545128.2 | XP_011543430.1 | 18 - 19 | 1973 | 69 | |
XM_011545129.2 | XP_011543431.1 | 21 - 22 | 2044 | 69 | |
XM_017017956.1 | XP_016873445.1 | 20 - 21 | 2060 | 69 | |
XM_017017957.1 | XP_016873446.1 | 19 - 20 | 2051 | 69 | |
GenBank mRNA | AB845669.1 | - | 20 - 21 | 2050 | 69 |
AK001123.1 | - | 10 - 11 | 893 | 69 | |
AK293088.1 | - | 20 - 21 | 2063 | 69 | |
AL833582.1 | - | 20 - 21 | 2099 | 69 | |
AY728143.1 | - | 20 - 21 | 2355 | 69 | |
BC027590.2 | - | 14 - 15 | 1229 | 69 | |
BC033036.2 | - | 18 - 19 | 2069 | 69 |