Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTDC1
Entrez Gene ID
79712
Gene Name
glycosyltransferase like domain containing 1
Gene Aliases
Hmat-Xa, mat-Xa
Chromosome Location
Chr.2: 143945787 - 144333092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006636.3 NP_001006637.1 11 - 12 1573 85
NM_001164629.3 NP_001158101.1 11 - 12 1558 85
NM_001284233.1 NP_001271162.1 8 - 9 1237 85
NM_001284234.1 NP_001271163.1 9 - 10 1273 85
NM_001284238.1 NP_001271167.1 13 - 14 1772 85
NM_024659.4 NP_078935.2 9 - 10 1285 85
XM_005263774.2 XP_005263831.1 12 - 13 1657 85
XM_005263782.2 XP_005263839.1 12 - 13 1673 85
XM_005263784.2 XP_005263841.1 10 - 11 1532 85
XM_011511843.2 XP_011510145.1 12 - 13 1706 85
XM_011511855.2 XP_011510157.1 10 - 11 1650 85
XM_011511856.2 XP_011510158.1 10 - 11 1432 85
XM_017004917.1 XP_016860406.1 16 - 17 2328 85
XM_017004918.1 XP_016860407.1 14 - 15 1920 85
XM_017004919.1 XP_016860408.1 12 - 13 1806 85
XM_017004920.1 XP_016860409.1 14 - 15 4962 85
XM_017004921.1 XP_016860410.1 13 - 14 1755 85
XM_017004922.1 XP_016860411.1 13 - 14 1932 85
XM_017004923.1 XP_016860412.1 12 - 13 1720 85
XM_017004924.1 XP_016860413.1 14 - 15 1916 85
XM_017004925.1 XP_016860414.1 11 - 12 1578 85
XM_017004926.1 XP_016860415.1 9 - 10 1509 85
XM_017004927.1 XP_016860416.1 11 - 12 1540 85
XM_017004928.1 XP_016860417.1 13 - 14 1773 85
XM_017004929.1 XP_016860418.1 11 - 12 1510 85
XM_017004930.1 XP_016860419.1 11 - 12 2161 85
XM_017004931.1 XP_016860420.1 11 - 12 1574 85
XM_017004932.1 XP_016860421.1 11 - 12 1544 85
XM_017004933.1 XP_016860422.1 11 - 12 1526 85
XM_017004934.1 XP_016860423.1 12 - 13 4733 85
XM_017004935.1 XP_016860424.1 13 - 14 2603 85
XM_017004936.1 XP_016860425.1 12 - 13 1780 85
XM_017004937.1 XP_016860426.1 9 - 10 1516 85
XM_017004938.1 XP_016860427.1 8 - 9 1404 85
XM_017004939.1 XP_016860428.1 11 - 12 1564 85
XM_017004940.1 XP_016860429.1 8 - 9 1365 85
XM_017004941.1 XP_016860430.1 7 - 8 1254 85
XM_017004942.1 XP_016860431.1 10 - 11 1411 85
XM_017004943.1 XP_016860432.1 10 - 11 2077 85
XM_017004944.1 XP_016860433.1 9 - 10 1300 85
XM_017004945.1 XP_016860434.1 11 - 12 1518 85
XM_017004946.1 XP_016860435.1 9 - 10 1255 85
XM_017004947.1 XP_016860436.1 9 - 10 1938 85
XM_017004948.1 XP_016860437.1 10 - 11 4478 85
XM_017004949.1 XP_016860438.1 8 - 9 1154 85
XM_017004950.1 XP_016860439.1 10 - 11 1201 85
XM_017004951.1 XP_016860440.1 10 - 11 1156 85
XM_017004952.1 XP_016860441.1 10 - 11 1600 85
XM_017004953.1 XP_016860442.1 10 - 11 1172 85
XM_017004954.1 XP_016860443.1 11 - 12 1260 85
XM_017004955.1 XP_016860444.1 13 - 14 1608 85
XM_017004956.1 XP_016860445.1 7 - 8 1011 85
XM_017004957.1 XP_016860446.1 9 - 10 1057 85
GenBank mRNA AB201269.1 - 8 - 9 1294 85
AB597184.1 - 8 - 9 1294 85
AK021815.1 - 5 - 6 749 85
AK091684.1 - 8 - 9 1241 85
AK291357.1 - 11 - 12 1488 85
AY281366.1 - 11 - 12 1573 85
BC017741.2 - 10 - 11 1325 85
BX647436.1 - 9 - 10 1255 85
BX647773.1 - 13 - 14 1711 85

View Full Product Details