The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020927.2 | NP_065978.1 | 1 - 2 | 354 | 66 |
GenBank mRNA | AB046796.1 | - | 1 - 2 | 324 | 66 |
AK290153.1 | - | 1 - 2 | 332 | 66 | |
BC033537.1 | - | 1 - 2 | 332 | 66 | |
HY132852.1 | - | 1 - 2 | 355 | 66 |