The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020927.2 | NP_065978.1 | 6 - 7 | 1000 | 101 |
GenBank mRNA | AB046796.1 | - | 6 - 7 | 970 | 101 |
AK290153.1 | - | 6 - 7 | 978 | 101 | |
AL834240.1 | - | 5 - 6 | 615 | 101 | |
BC033537.1 | - | 6 - 7 | 978 | 101 |