Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKS1B
Entrez Gene ID
56899
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1B
Gene Aliases
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
Chromosome Location
Chr.12: 98729904 - 99984727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204065.1 NP_001190994.1 8 - 9 922 123
NM_001204066.1 NP_001190995.1 9 - 10 789 123
NM_001204067.1 NP_001190996.1 9 - 10 883 123
NM_001204068.1 NP_001190997.1 9 - 10 1346 123
NM_001204069.1 NP_001190998.1 9 - 10 1238 123
NM_001204070.1 NP_001190999.1 8 - 9 1166 123
NM_001204079.1 NP_001191008.1 8 - 9 925 123
NM_001204080.1 NP_001191009.1 7 - 8 850 123
NM_001204081.1 NP_001191010.1 7 - 8 850 123
NM_020140.3 NP_064525.1 8 - 9 1166 123
NM_152788.4 NP_690001.3 23 - 24 3778 123
NM_181670.3 NP_858056.2 10 - 11 1418 123
XM_005269028.4 XP_005269085.1 23 - 24 3389 123
XM_005269029.4 XP_005269086.1 23 - 24 3349 123
XM_005269032.3 XP_005269089.1 7 - 8 714 123
XM_006719504.3 XP_006719567.1 24 - 25 3923 123
XM_006719505.3 XP_006719568.1 24 - 25 3926 123
XM_006719506.3 XP_006719569.1 24 - 25 3923 123
XM_006719507.3 XP_006719570.1 24 - 25 3926 123
XM_006719508.3 XP_006719571.1 24 - 25 3926 123
XM_006719509.3 XP_006719572.1 23 - 24 3851 123
XM_006719510.3 XP_006719573.1 23 - 24 3851 123
XM_006719512.3 XP_006719575.1 22 - 23 3671 123
XM_006719513.3 XP_006719576.1 22 - 23 3671 123
XM_006719514.3 XP_006719577.1 22 - 23 3659 123
XM_011538571.2 XP_011536873.1 24 - 25 3824 123
XM_017019651.1 XP_016875140.1 24 - 25 3926 123
XM_017019652.1 XP_016875141.1 24 - 25 3914 123
XM_017019653.1 XP_016875142.1 24 - 25 3923 123
XM_017019654.1 XP_016875143.1 24 - 25 3911 123
XM_017019655.1 XP_016875144.1 24 - 25 3923 123
XM_017019656.1 XP_016875145.1 24 - 25 3821 123
XM_017019657.1 XP_016875146.1 23 - 24 3749 123
XM_017019658.1 XP_016875147.1 22 - 23 3671 123
XM_017019659.1 XP_016875148.1 10 - 11 1417 123
XM_017019660.1 XP_016875149.1 9 - 10 1342 123
XM_017019661.1 XP_016875150.1 11 - 12 1128 123
XM_017019662.1 XP_016875151.1 11 - 12 1261 123
XM_017019663.1 XP_016875152.1 10 - 11 1312 123
XM_017019664.1 XP_016875153.1 10 - 11 998 123
XM_017019665.1 XP_016875154.1 8 - 9 1162 123
GenBank mRNA AF145204.1 - 17 - 18 2482 123
AF164792.1 - 8 - 9 1099 123
AK289768.1 - 10 - 11 1383 123
AK294191.1 - 9 - 10 883 123
AK294994.1 - 7 - 8 722 123
AK295988.1 - 8 - 9 895 123
AK315954.1 - 8 - 9 777 123
AM980945.1 - 8 - 9 781 123
AY281131.1 - 23 - 24 3364 123
AY281132.1 - 9 - 10 1039 123
AY283057.2 - 15 - 16 2215 123
AY356353.1 - 7 - 8 679 123
AY620824.1 - 14 - 15 2089 123
AY753193.1 - 8 - 9 859 123
BC026313.1 - 10 - 11 1313 123
BC068451.1 - 9 - 10 789 123
BC142669.1 - 8 - 9 1113 123
BC150204.1 - 9 - 10 1185 123

View Full Product Details