Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFT2D3
Entrez Gene ID
84826
Gene Name
SFT2 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 127701023 - 127703833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032740.3 NP_116129.3 1 - 1 2676 142
GenBank mRNA AK074549.1 - 1 - 1 2191 142
BC006808.1 - 1 - 1 1033 142
BC063637.1 - 1 - 1 1144 142
BC110324.1 - 1 - 1 1146 142

View Full Product Details