The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170723.1 | NP_001164194.1 | 1 - 2 | 155 | 143 |
NM_001170724.1 | NP_001164195.1 | 1 - 2 | 155 | 143 | |
NM_032126.4 | NP_115502.2 | 1 - 2 | 155 | 143 | |
GenBank mRNA | AK093269.1 | - | 1 - 2 | 102 | 143 |
BI461686.1 | - | 1 - 2 | 120 | 143 | |
BI825610.1 | - | 1 - 2 | 89 | 143 |