The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170722.1 | NP_001164193.1 | 2 - 3 | 218 | 163 |
NM_001170723.1 | NP_001164194.1 | 2 - 3 | 194 | 139 | |
NM_001170724.1 | NP_001164195.1 | 2 - 3 | 194 | 139 | |
NM_032126.4 | NP_115502.2 | 2 - 3 | 194 | 139 | |
GenBank mRNA | AK093269.1 | - | 2 - 3 | 141 | 139 |
AL136694.1 | - | 2 - 3 | 198 | 139 | |
BC034972.2 | - | 2 - 3 | 151 | 163 | |
BI461686.1 | - | 2 - 3 | 159 | 139 | |
BI825610.1 | - | 2 - 3 | 128 | 139 |