The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032126.4 | NP_115502.2 | 7 - 8 | 656 | 137 |
GenBank mRNA | AL136694.1 | - | 7 - 8 | 660 | 137 |