The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026716.2 | 1 - 2 | 139 | 68 | |
NR_104095.1 | 1 - 2 | 139 | 68 | ||
NR_104096.1 | 1 - 2 | 139 | 68 | ||
NR_104097.1 | 1 - 2 | 139 | 68 | ||
NR_136268.1 | 1 - 2 | 139 | 68 | ||
GenBank mRNA | BC171827.1 | - | 1 - 2 | 139 | 68 |
BC171832.1 | - | 1 - 2 | 139 | 68 | |
BC171838.1 | - | 1 - 2 | 139 | 68 | |
DQ224421.1 | - | 1 - 2 | 40 | 68 |