The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025104.1 | NP_001020275.1 | 3 - 4 | 130 | 74 |
NM_001890.1 | NP_001881.1 | 3 - 4 | 130 | 74 | |
XM_006714089.2 | XP_006714152.1 | 3 - 4 | 149 | 74 | |
XM_006714090.2 | XP_006714153.1 | 3 - 4 | 149 | 74 | |
XM_006714091.2 | XP_006714154.1 | 3 - 4 | 149 | 74 | |
GenBank mRNA | BC128227.1 | - | 3 - 4 | 119 | 74 |
BC128228.1 | - | 3 - 4 | 119 | 74 | |
BG217918.1 | - | 2 - 3 | 92 | 74 | |
DQ064604.1 | - | 2 - 3 | 81 | 74 | |
U23157.1 | - | 3 - 4 | 109 | 74 | |
X78416.1 | - | 3 - 4 | 130 | 74 | |
X98084.1 | - | 2 - 3 | 58 | 74 |