The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017682.2 | NP_060152.2 | 2 - 3 | 249 | 78 |
XM_005259963.3 | XP_005260020.1 | 3 - 4 | 449 | 78 | |
GenBank mRNA | AF440756.1 | - | 2 - 3 | 249 | 78 |
AK000139.1 | - | 2 - 3 | 184 | 78 | |
AK225232.1 | - | 2 - 3 | 184 | 78 | |
AY515705.1 | - | 2 - 3 | 249 | 78 |