Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BEST2
Entrez Gene ID
54831
Gene Name
bestrophin 2
Gene Aliases
VMD2L1
Chromosome Location
Chr.19: 12751691 - 12758458 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017682.2 NP_060152.2 4 - 5 643 137
XM_005259963.3 XP_005260020.1 5 - 6 843 137
GenBank mRNA AF440756.1 - 4 - 5 643 137
AK000139.1 - 4 - 5 578 137
AK225232.1 - 4 - 5 578 137
AY515705.1 - 4 - 5 643 137

View Full Product Details