The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181453.3 | NP_852118.1 | 8 - 9 | 3118 | 93 |
NR_028063.1 | 7 - 8 | 3033 | 93 | ||
XM_006712870.3 | XP_006712933.1 | 8 - 9 | 3687 | 93 | |
XM_006712871.1 | XP_006712934.1 | 7 - 8 | 2941 | 93 | |
XM_011512213.2 | XP_011510515.1 | 7 - 8 | 2934 | 93 | |
GenBank mRNA | AB002334.1 | - | 7 - 8 | 2914 | 93 |
BC146789.1 | - | 7 - 8 | 2914 | 93 |