Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TATDN2
Entrez Gene ID
9797
Gene Name
TatD DNase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 10248493 - 10281222 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014760.3 NP_055575.3 4 - 5 2411 123
GenBank mRNA AK289794.1 - 4 - 5 2132 123
BC090935.1 - 4 - 5 1992 123
BC101770.1 - 4 - 5 1900 123
BC101776.1 - 4 - 5 1901 123
D86972.1 - 5 - 6 2228 123

View Full Product Details