Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM49B
Entrez Gene ID
51571
Gene Name
family with sequence similarity 49 member B
Gene Aliases
BM-009, L1
Chromosome Location
Chr.8: 129839593 - 130016697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256763.1 NP_001243692.1 7 - 8 839 115
NM_016623.4 NP_057707.3 6 - 7 752 115
NR_046359.1 9 - 10 1105 115
NR_046360.1 9 - 10 1015 115
NR_046361.1 8 - 9 1005 115
XM_005250966.3 XP_005251023.1 5 - 6 608 115
XM_011517108.2 XP_011515410.1 8 - 9 901 115
XM_011517109.2 XP_011515411.1 8 - 9 673 115
XM_011517110.2 XP_011515412.1 7 - 8 813 115
XM_011517111.2 XP_011515413.1 7 - 8 631 115
XM_011517112.2 XP_011515414.1 9 - 10 856 115
XM_011517113.2 XP_011515415.1 8 - 9 867 115
XM_011517114.2 XP_011515416.1 8 - 9 773 115
XM_011517115.2 XP_011515417.1 7 - 8 780 115
XM_011517116.2 XP_011515418.1 8 - 9 814 115
XM_011517117.2 XP_011515419.1 7 - 8 783 115
XM_011517118.2 XP_011515420.1 7 - 8 727 115
XM_011517120.2 XP_011515422.1 6 - 7 726 115
XM_011517121.2 XP_011515423.1 9 - 10 738 115
XM_011517122.2 XP_011515424.1 7 - 8 817 115
XM_011517123.2 XP_011515425.1 7 - 8 590 115
XM_011517124.2 XP_011515426.1 6 - 7 705 115
XM_011517125.2 XP_011515427.1 6 - 7 644 115
XM_017013539.1 XP_016869028.1 9 - 10 860 115
XM_017013540.1 XP_016869029.1 9 - 10 719 115
XM_017013541.1 XP_016869030.1 7 - 8 847 115
XM_017013542.1 XP_016869031.1 9 - 10 897 115
XM_017013543.1 XP_016869032.1 9 - 10 997 115
XM_017013544.1 XP_016869033.1 9 - 10 779 115
XM_017013545.1 XP_016869034.1 10 - 11 1043 115
XM_017013546.1 XP_016869035.1 8 - 9 810 115
XM_017013547.1 XP_016869036.1 10 - 11 902 115
XM_017013548.1 XP_016869037.1 8 - 9 906 115
XM_017013549.1 XP_016869038.1 8 - 9 769 115
XM_017013550.1 XP_016869039.1 7 - 8 820 115
XM_017013551.1 XP_016869040.1 9 - 10 815 115
XM_017013552.1 XP_016869041.1 9 - 10 860 115
XM_017013553.1 XP_016869042.1 8 - 9 673 115
XM_017013554.1 XP_016869043.1 10 - 11 885 115
XM_017013555.1 XP_016869044.1 7 - 8 840 115
XM_017013556.1 XP_016869045.1 7 - 8 586 115
XM_017013557.1 XP_016869046.1 8 - 9 632 115
XM_017013558.1 XP_016869047.1 8 - 9 777 115
XM_017013559.1 XP_016869048.1 8 - 9 636 115
XM_017013560.1 XP_016869049.1 6 - 7 751 115
XM_017013561.1 XP_016869050.1 6 - 7 503 115
XM_017013562.1 XP_016869051.1 7 - 8 690 115
GenBank mRNA AK001226.1 - 2 - 3 258 115
AK002059.1 - 9 - 10 815 115
AK130372.1 - 6 - 7 639 115
BC003599.1 - 7 - 8 618 115
BC017297.2 - 9 - 10 987 115
CR749628.1 - 7 - 8 585 115

View Full Product Details