Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP6
Entrez Gene ID
4135
Gene Name
microtubule associated protein 6
Gene Aliases
MAP6-N, MTAP6, N-STOP, STOP
Chromosome Location
Chr.11: 75586914 - 75671108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006718556.3 XP_006718619.1 1 - 1 2621 96
XM_011545039.2 XP_011543341.1 1 - 1 2622 96
XM_017017755.1 XP_016873244.1 1 - 1 2521 96


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR740965.1 - 1 Chr. 11 - 75668372 - 75668713 [+] on Build GRCh38 96
GenBankmRNA DR763219.1 - 1 39 Chr. 11 - 75667738 - 75668491 [-] on Build GRCh38 96
GenBankmRNA BF363002.1 - 1 Chr. 11 - 75668270 - 75668495 [+] on Build GRCh38 96
GenBankmRNA DR003017.1 - 1 222 Chr. 11 - 75668094 - 75668665 [-] on Build GRCh38 96
GenBankmRNA DA743021.1 - 1 Chr. 11 - 75668216 - 75668730 [+] on Build GRCh38 96
GenBankmRNA CR609646.1 - 1 Chr. 11 - 75667664 - 75669120 [+] on Build GRCh38 96
GenBankmRNA BX339321.2 - 1 Chr. 11 - 75667696 - 75668656 [+] on Build GRCh38 96
GenBankmRNA HY121308.1 - 1 148 Chr. 11 - 75668268 - 75668597 [-] on Build GRCh38 96
GenBankmRNA DA339628.1 - 1 120 Chr. 11 - 75608080 - 75668572 [-] on Build GRCh38 96


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream serpin family H member 1 SERPINH1 AsTP3, CBP1, CBP2, HSP47, OI10, PIG14, PPROM, RA-A47, SERPINH2, gp46 -94.9kb Anti-sense Chr. - 75562056 - 75572804 on Build GRCh38
Downstream monoacylglycerol O-acyltransferase 2 MOGAT2 DGAT2L5, DGAT2L5., MGAT2, hDC5 49.3kb Anti-sense Chr. - 75717819 - 75732958 on Build GRCh38

View Full Product Details