The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194946.1 | NP_001181875.1 | 24 - 25 | 3716 | 67 |
NM_001194947.1 | NP_001181876.1 | 23 - 24 | 3659 | 67 | |
NM_001291157.1 | NP_001278086.1 | 23 - 24 | 3748 | 67 | |
NM_004953.4 | NP_004944.3 | 16 - 17 | 3207 | 67 | |
NM_182917.4 | NP_886553.3 | 22 - 23 | 3843 | 67 | |
NM_198241.2 | NP_937884.1 | 23 - 24 | 3695 | 67 | |
NM_198242.2 | NP_937885.1 | 19 - 20 | 3280 | 67 | |
NM_198244.2 | NP_937887.1 | 20 - 21 | 3457 | 67 | |
GenBank mRNA | AB210013.1 | - | 22 - 23 | 3834 | 67 |
AF104913.1 | - | 20 - 21 | 3319 | 67 | |
AF281070.1 | - | 22 - 23 | 3701 | 67 | |
AK226160.1 | - | 23 - 24 | 3629 | 67 | |
AK294567.1 | - | 21 - 22 | 3326 | 67 | |
AK299763.1 | - | 20 - 21 | 3385 | 67 | |
AL832754.1 | - | 19 - 20 | 3169 | 67 | |
AY082886.1 | - | 22 - 23 | 3701 | 67 | |
BC007788.2 | - | 5 - 6 | 563 | 67 | |
BC082764.1 | - | 16 - 17 | 2853 | 67 | |
BC140892.1 | - | 24 - 25 | 3558 | 67 | |
BC140896.1 | - | 23 - 24 | 3501 | 67 | |
BT007282.1 | - | 5 - 6 | 562 | 67 | |
BX647812.1 | - | 23 - 24 | 3608 | 67 | |
CX165113.1 | - | 2 - 3 | 148 | 67 | |
D12686.1 | - | 16 - 17 | 3198 | 67 |