The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024602.5 | NP_078878.3 | 9 - 10 | 1417 | 59 |
XM_011542140.2 | XP_011540442.1 | 7 - 8 | 814 | 59 | |
GenBank mRNA | AK056545.1 | - | 7 - 8 | 768 | 59 |
AK056873.1 | - | 1 - 2 | 311 | 59 | |
AK096462.1 | - | 9 - 10 | 1324 | 59 | |
AK301124.1 | - | 8 - 9 | 1005 | 59 | |
BC126352.1 | - | 6 - 7 | 694 | 59 | |
BC130470.1 | - | 6 - 7 | 694 | 59 |