The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006224.3 | NP_006215.1 | 8 - 9 | 791 | 105 |
XM_011523939.1 | XP_011522241.1 | 7 - 8 | 703 | 105 | |
XM_011523940.1 | XP_011522242.1 | 8 - 9 | 1213 | 105 | |
GenBank mRNA | AK303986.1 | - | 8 - 9 | 561 | 105 |
AL542612.3 | - | 8 - 9 | 789 | 105 | |
BC027177.1 | - | 6 - 7 | 638 | 105 | |
BC045108.1 | - | 8 - 9 | 766 | 105 | |
BC068538.1 | - | 8 - 9 | 674 | 105 | |
BC082976.1 | - | 8 - 9 | 706 | 105 | |
D30036.1 | - | 8 - 9 | 743 | 105 | |
EU794647.1 | - | 8 - 9 | 766 | 105 | |
M73704.1 | - | 8 - 9 | 751 | 105 |