Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC8A
Entrez Gene ID
55220
Gene Name
kelch domain containing 8A
Gene Aliases
-
Chromosome Location
Chr.1: 205336065 - 205357090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271863.1 NP_001258792.1 8 - 9 1728 112
NM_001271864.1 NP_001258793.1 6 - 7 1188 112
NM_001271865.1 NP_001258794.1 7 - 8 1403 112
NM_018203.2 NP_060673.1 5 - 6 1516 112
GenBank mRNA AK001610.1 - 5 - 6 1427 112
AK096590.1 - 8 - 9 1677 112
AK299227.1 - 4 - 5 739 112
BC036076.1 - 5 - 6 1097 112

View Full Product Details