Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
Chromosome Location
Chr.12: 103587250 - 103766724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017564.9 NP_060034.9 41 - 42 4598 64
XM_011538537.2 XP_011536839.1 41 - 42 4639 64
XM_011538538.2 XP_011536840.1 41 - 42 4639 64
XM_011538539.2 XP_011536841.1 27 - 28 2842 64
XM_011538541.2 XP_011536843.1 19 - 20 1949 64
XM_011538542.2 XP_011536844.1 8 - 9 758 64
XM_017019585.1 XP_016875074.1 41 - 42 4639 64
GenBank mRNA AB052958.1 - 41 - 42 4594 64
AJ295695.1 - 41 - 42 4598 64
AK024503.1 - 9 - 10 940 64
AK074051.1 - 19 - 20 1969 64
AK160380.1 - 40 - 41 4311 64
AL133021.1 - 1 - 2 80 64
AY227444.1 - 10 - 11 1007 64
AY311388.1 - 41 - 42 4616 64

View Full Product Details