Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAB2
Entrez Gene ID
55576
Gene Name
stabilin 2
Gene Aliases
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1
Chromosome Location
Chr.12: 103587250 - 103766724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017564.9 NP_060034.9 48 - 49 5269 151
XM_011538537.2 XP_011536839.1 48 - 49 5310 151
XM_011538538.2 XP_011536840.1 48 - 49 5310 151
XM_011538539.2 XP_011536841.1 34 - 35 3513 151
XM_011538541.2 XP_011536843.1 26 - 27 2620 151
XM_011538542.2 XP_011536844.1 15 - 16 1429 151
XM_017019585.1 XP_016875074.1 48 - 49 5310 151
GenBank mRNA AB052958.1 - 48 - 49 5265 151
AF160476.1 - 3 - 4 291 151
AJ295695.1 - 48 - 49 5269 151
AK024503.1 - 16 - 17 1611 151
AK074051.1 - 26 - 27 2640 151
AK160380.1 - 47 - 48 4987 151
AL133021.1 - 8 - 9 751 151
AY227444.1 - 17 - 18 1678 151
AY311388.1 - 48 - 49 5287 151

View Full Product Details