The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199573.1 | NP_001186502.1 | 4 - 5 | 1114 | 117 |
NM_006074.4 | NP_006065.2 | 4 - 5 | 1126 | 117 | |
GenBank mRNA | AK290434.1 | - | 4 - 5 | 856 | 117 |
AK298715.1 | - | 5 - 6 | 806 | 117 | |
AK298934.1 | - | 4 - 5 | 680 | 117 | |
AK301192.1 | - | 3 - 4 | 597 | 117 | |
AK308796.1 | - | 3 - 4 | 746 | 117 | |
AL360134.1 | - | 2 - 3 | 284 | 117 | |
BC022281.1 | - | 4 - 5 | 933 | 117 | |
BC035582.1 | - | 4 - 5 | 899 | 117 | |
HQ842635.1 | - | 3 - 4 | 747 | 117 | |
X82200.1 | - | 4 - 5 | 857 | 117 |