The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199573.1 | NP_001186502.1 | 6 - 7 | 1248 | 62 |
NM_006074.4 | NP_006065.2 | 6 - 7 | 1260 | 62 | |
GenBank mRNA | AK290434.1 | - | 6 - 7 | 990 | 62 |
AK298715.1 | - | 7 - 8 | 940 | 62 | |
AK298934.1 | - | 6 - 7 | 814 | 62 | |
AK301192.1 | - | 5 - 6 | 731 | 62 | |
AK310098.1 | - | 5 - 6 | 744 | 62 | |
AL360134.1 | - | 4 - 5 | 418 | 62 | |
AM040752.1 | - | 3 - 4 | 208 | 62 | |
BC022281.1 | - | 6 - 7 | 1067 | 62 | |
BC035582.1 | - | 6 - 7 | 1033 | 62 | |
HQ842635.1 | - | 5 - 6 | 881 | 62 | |
X82200.1 | - | 6 - 7 | 991 | 62 |