The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008723.1 | NP_001008723.1 | 12 - 13 | 1945 | 106 |
XM_011539377.2 | XP_011537679.1 | 12 - 13 | 2664 | 106 | |
XM_017015787.1 | XP_016871276.1 | 12 - 13 | 2316 | 106 | |
XM_017015788.1 | XP_016871277.1 | 12 - 13 | 2847 | 106 | |
GenBank mRNA | AK296463.1 | - | 12 - 13 | 1938 | 106 |
BC036225.1 | - | 12 - 13 | 1947 | 106 | |
BC109126.2 | - | 12 - 13 | 1899 | 106 | |
BX641120.1 | - | 12 - 13 | 1947 | 106 |