The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011649.2 | NP_001011649.1 | 33 - 34 | 5256 | 76 |
NM_001272039.1 | NP_001258968.1 | 31 - 32 | 4803 | 76 | |
NM_018249.5 | NP_060719.4 | 34 - 35 | 5493 | 76 | |
NR_073554.1 | 34 - 35 | 5568 | 76 | ||
NR_073555.1 | 34 - 35 | 5491 | 76 | ||
NR_073556.1 | 35 - 36 | 5700 | 76 | ||
NR_073557.1 | 34 - 35 | 5573 | 76 | ||
NR_073558.1 | 34 - 35 | 5570 | 76 | ||
XM_006717182.1 | XP_006717245.1 | 33 - 34 | 5386 | 76 | |
XM_006717185.1 | XP_006717248.1 | 31 - 32 | 4795 | 76 | |
XM_011518860.1 | XP_011517162.1 | 34 - 35 | 5479 | 76 | |
XM_017014921.1 | XP_016870410.1 | 33 - 34 | 5383 | 76 | |
XM_017014922.1 | XP_016870411.1 | 28 - 29 | 4711 | 76 | |
XM_017014923.1 | XP_016870412.1 | 30 - 31 | 4558 | 76 | |
XM_017014924.1 | XP_016870413.1 | 24 - 25 | 4230 | 76 | |
GenBank mRNA | AB046853.2 | - | 33 - 34 | 5225 | 76 |
AF087969.1 | - | 1 - 2 | 205 | 76 | |
AF448860.1 | - | 34 - 35 | 5443 | 76 | |
AK001729.1 | - | 12 - 13 | 2331 | 76 | |
AK025867.1 | - | 6 - 7 | 823 | 76 | |
AK027636.1 | - | 13 - 14 | 2457 | 76 | |
AK122913.1 | - | 34 - 35 | 5482 | 76 | |
AL133161.1 | - | 18 - 19 | 3483 | 76 | |
BC004526.2 | - | 7 - 8 | 1102 | 76 | |
BC136275.1 | - | 34 - 35 | 5325 | 76 | |
BC143762.1 | - | 33 - 34 | 5091 | 76 | |
BC146782.1 | - | 33 - 34 | 5225 | 76 | |
BX537421.1 | - | 34 - 35 | 5478 | 76 | |
BX537708.1 | - | 31 - 32 | 4792 | 76 | |
BX537759.1 | - | 33 - 34 | 5244 | 76 | |
CR936622.1 | - | 27 - 28 | 4623 | 76 | |
CR936717.1 | - | 27 - 28 | 4623 | 76 | |
KP294527.1 | - | 34 - 35 | 5493 | 76 |