The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138353.2 | NP_612362.2 | 4 - 5 | 480 | 86 |
GenBank mRNA | AK093072.1 | - | 4 - 5 | 497 | 86 |
BC002926.2 | - | 2 - 3 | 234 | 86 | |
BC013280.2 | - | 2 - 3 | 235 | 86 | |
BC018894.2 | - | 2 - 3 | 234 | 86 | |
BC018895.2 | - | 2 - 3 | 234 | 86 | |
BC080575.1 | - | 4 - 5 | 480 | 86 |