The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 3 - 4 | 384 | 78 |
GenBank mRNA | AY826979.1 | - | 3 - 4 | 373 | 78 |
BC127666.1 | - | 3 - 4 | 384 | 78 | |
BC127667.1 | - | 3 - 4 | 388 | 78 | |
J02703.1 | - | 3 - 4 | 384 | 78 | |
KC120777.1 | - | 3 - 4 | 384 | 78 | |
M20311.1 | - | 3 - 4 | 380 | 78 | |
M35999.1 | - | 3 - 4 | 382 | 78 | |
U95204.1 | - | 3 - 4 | 364 | 78 |