The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004555.3 | NP_004546.1 | 4 - 5 | 1914 | 122 |
NM_173163.2 | NP_775186.1 | 4 - 5 | 1914 | 122 | |
NM_173165.2 | NP_775188.1 | 4 - 5 | 1914 | 122 | |
GenBank mRNA | AB209824.1 | - | 2 - 3 | 541 | 122 |
AK290370.1 | - | 4 - 5 | 1654 | 122 | |
BC001050.2 | - | 4 - 5 | 1632 | 122 | |
EU887605.1 | - | 4 - 5 | 1598 | 122 | |
EU887606.1 | - | 4 - 5 | 1598 | 122 | |
EU887607.1 | - | 4 - 5 | 1598 | 122 | |
EU887608.1 | - | 4 - 5 | 1925 | 122 | |
EU887609.1 | - | 4 - 5 | 1925 | 122 | |
EU887610.1 | - | 4 - 5 | 1925 | 122 | |
EU887611.1 | - | 4 - 5 | 1643 | 122 | |
EU887612.1 | - | 4 - 5 | 1643 | 122 | |
EU887613.1 | - | 4 - 5 | 1643 | 122 | |
EU887614.1 | - | 4 - 5 | 1551 | 122 | |
EU887615.1 | - | 4 - 5 | 1551 | 122 | |
EU887616.1 | - | 4 - 5 | 1551 | 122 | |
EU887617.1 | - | 4 - 5 | 1556 | 122 | |
EU887618.1 | - | 4 - 5 | 1556 | 122 | |
EU887619.1 | - | 4 - 5 | 1556 | 122 | |
EU887620.1 | - | 4 - 5 | 1593 | 122 | |
EU887621.1 | - | 4 - 5 | 1593 | 122 | |
EU887622.1 | - | 4 - 5 | 1593 | 122 | |
EU887623.1 | - | 4 - 5 | 1997 | 122 | |
EU887624.1 | - | 4 - 5 | 1997 | 122 | |
EU887625.1 | - | 4 - 5 | 1997 | 122 | |
L41067.1 | - | 4 - 5 | 1808 | 122 | |
U14510.1 | - | 4 - 5 | 1622 | 122 | |
U85428.1 | - | 4 - 5 | 1622 | 122 | |
U85429.1 | - | 4 - 5 | 1622 | 122 | |
U85430.1 | - | 4 - 5 | 1622 | 122 |