The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288772.1 | NP_001275701.1 | 27 - 28 | 3900 | 74 |
NM_001288774.1 | NP_001275703.1 | 27 - 28 | 3893 | 74 | |
NM_004570.5 | NP_004561.3 | 26 - 27 | 3777 | 74 | |
XM_011520696.2 | XP_011518998.1 | 27 - 28 | 4028 | 74 | |
XM_011520697.2 | XP_011518999.1 | 27 - 28 | 4027 | 74 | |
XM_017019470.1 | XP_016874959.1 | 27 - 28 | 4028 | 74 | |
XM_017019471.1 | XP_016874960.1 | 27 - 28 | 3965 | 74 | |
XM_017019472.1 | XP_016874961.1 | 28 - 29 | 4193 | 74 | |
XM_017019473.1 | XP_016874962.1 | 27 - 28 | 5034 | 74 | |
XM_017019474.1 | XP_016874963.1 | 26 - 27 | 3905 | 74 | |
XM_017019475.1 | XP_016874964.1 | 27 - 28 | 4027 | 74 | |
XM_017019476.1 | XP_016874965.1 | 25 - 26 | 3774 | 74 | |
XM_017019477.1 | XP_016874966.1 | 26 - 27 | 3215 | 74 | |
XM_017019479.1 | XP_016874968.1 | 14 - 15 | 1939 | 74 | |
GenBank mRNA | BC130277.1 | - | 25 - 26 | 3696 | 74 |
BC144133.1 | - | 26 - 27 | 3815 | 74 | |
BC144134.1 | - | 26 - 27 | 3819 | 74 |