Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
Chromosome Location
Chr.12: 13537337 - 13982012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000834.3 NP_000825.2 6 - 7 1676 75
XM_011520628.2 XP_011518930.1 7 - 8 2469 75
XM_011520629.2 XP_011518931.1 8 - 9 2276 75
XM_017019219.1 XP_016874708.1 7 - 8 2024 75
GenBank mRNA AB208850.1 - 3 - 4 455 75
BC113618.1 - 6 - 7 1575 75
BC113620.1 - 6 - 7 1575 75
U11287.1 - 6 - 7 1707 75
U88963.1 - 6 - 7 1676 75
U90278.1 - 6 - 7 1706 75

View Full Product Details