The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031488.4 | NP_113676.2 | 13 - 14 | 1644 | 64 |
XM_011530420.2 | XP_011528722.1 | 12 - 13 | 1627 | 64 | |
XM_011530421.2 | XP_011528723.1 | 13 - 14 | 1713 | 64 | |
XM_017028976.1 | XP_016884465.1 | 11 - 12 | 1421 | 64 | |
XM_017028977.1 | XP_016884466.1 | 12 - 13 | 1940 | 64 | |
XM_017028978.1 | XP_016884467.1 | 13 - 14 | 1895 | 64 | |
XM_017028979.1 | XP_016884468.1 | 13 - 14 | 1840 | 64 | |
XM_017028980.1 | XP_016884469.1 | 12 - 13 | 1527 | 64 | |
GenBank mRNA | AJ305226.1 | - | 13 - 14 | 1629 | 64 |
AJ305227.1 | - | 13 - 14 | 1629 | 64 | |
AK056443.1 | - | 13 - 14 | 1641 | 64 | |
AK074091.1 | - | 12 - 13 | 3848 | 64 | |
AK097052.1 | - | 13 - 14 | 1713 | 64 | |
AL136564.1 | - | 13 - 14 | 1644 | 64 | |
BC017191.2 | - | 13 - 14 | 1633 | 64 | |
CR456482.1 | - | 13 - 14 | 1611 | 64 | |
KU178799.1 | - | 13 - 14 | 1590 | 64 | |
KU178800.1 | - | 13 - 14 | 1590 | 64 |