Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN2
Entrez Gene ID
2199
Gene Name
fibulin 2
Gene Aliases
-
Chromosome Location
Chr.3: 13549125 - 13638422 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004019.1 NP_001004019.1 11 - 12 2697 69
NM_001165035.1 NP_001158507.1 11 - 12 2954 69
NM_001998.2 NP_001989.2 10 - 11 2556 69
XM_006713026.2 XP_006713089.1 10 - 11 2606 69
GenBank mRNA AK129883.1 - - 2041 69
AK300913.1 - 6 - 7 707 69
AK304777.1 - 10 - 11 2656 69
AK304827.1 - 11 - 12 2954 69
AL050095.1 - 5 - 6 608 69
AY130459.1 - 11 - 12 2641 69
BC051690.1 - 10 - 11 2519 69
X82494.1 - 10 - 11 2500 69

View Full Product Details