The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319842.1 | NP_001306771.1 | 13 - 14 | 3133 | 121 |
NM_001319843.1 | NP_001306772.1 | 14 - 15 | 3216 | 121 | |
NM_023925.4 | NP_076414.2 | 13 - 14 | 3069 | 121 | |
NM_032156.4 | NP_115532.3 | 14 - 15 | 3216 | 121 | |
XM_006719140.3 | XP_006719203.1 | 13 - 14 | 3234 | 121 | |
XM_017019852.1 | XP_016875341.1 | 15 - 16 | 2434 | 121 | |
XM_017019853.1 | XP_016875342.1 | 15 - 16 | 2504 | 121 | |
XM_017019854.1 | XP_016875343.1 | 12 - 13 | 3087 | 121 | |
XM_017019857.1 | XP_016875346.1 | 14 - 15 | 2329 | 121 | |
XM_017019858.1 | XP_016875347.1 | 14 - 15 | 2356 | 121 | |
XM_017019861.1 | XP_016875350.1 | 13 - 14 | 2209 | 121 | |
XM_017019862.1 | XP_016875351.1 | 14 - 15 | 3339 | 121 | |
XM_017019868.1 | XP_016875357.1 | 15 - 16 | 2434 | 121 | |
XM_017019869.1 | XP_016875358.1 | 15 - 16 | 2461 | 121 | |
XM_017019874.1 | XP_016875363.1 | 14 - 15 | 2351 | 121 | |
XM_017019878.1 | XP_016875367.1 | 11 - 12 | 1857 | 121 | |
XM_017019881.1 | XP_016875370.1 | 14 - 15 | 2351 | 121 | |
GenBank mRNA | AL834212.1 | - | 11 - 12 | 1703 | 121 |
AY074490.1 | - | 13 - 14 | 3069 | 121 | |
AY074491.1 | - | 13 - 14 | 3133 | 121 | |
BC117672.1 | - | 14 - 15 | 3013 | 121 |