Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location
Chr.12: 30709552 - 30754994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002259.2 NP_001002259.1 17 - 18 3570 58
NM_001319842.1 NP_001306771.1 16 - 17 3484 58
NM_001319843.1 NP_001306772.1 17 - 18 3567 58
NM_001319844.1 NP_001306773.1 17 - 18 2720 58
NM_001319845.1 NP_001306774.1 18 - 19 2825 58
NM_001319846.1 NP_001306775.1 18 - 19 2815 58
NM_023925.4 NP_076414.2 16 - 17 3420 58
XM_006719138.3 XP_006719201.1 17 - 18 3630 58
XM_006719139.3 XP_006719202.1 16 - 17 3588 58
XM_006719140.3 XP_006719203.1 16 - 17 3585 58
XM_006719142.3 XP_006719205.1 16 - 17 3483 58
XM_006719144.3 XP_006719207.1 15 - 16 3441 58
XM_017019851.1 XP_016875340.1 16 - 17 3525 58
XM_017019852.1 XP_016875341.1 18 - 19 2785 58
XM_017019853.1 XP_016875342.1 18 - 19 2855 58
XM_017019854.1 XP_016875343.1 15 - 16 3438 58
XM_017019855.1 XP_016875344.1 18 - 19 2752 58
XM_017019856.1 XP_016875345.1 17 - 18 2710 58
XM_017019857.1 XP_016875346.1 17 - 18 2680 58
XM_017019858.1 XP_016875347.1 17 - 18 2707 58
XM_017019859.1 XP_016875348.1 16 - 17 2536 58
XM_017019860.1 XP_016875349.1 18 - 19 3737 58
XM_017019861.1 XP_016875350.1 16 - 17 2560 58
XM_017019862.1 XP_016875351.1 18 - 19 3734 58
XM_017019874.1 XP_016875363.1 17 - 18 2702 58
XM_017019876.1 XP_016875365.1 15 - 16 2253 58
XM_017019877.1 XP_016875366.1 14 - 15 2211 58
XM_017019878.1 XP_016875367.1 14 - 15 2208 58
XM_017019879.1 XP_016875368.1 16 - 17 2537 58
XM_017019880.1 XP_016875369.1 13 - 14 2001 58
GenBank mRNA AF525416.1 - 9 - 10 1081 58
AK024247.1 - 4 - 5 2537 58
AK026222.1 - 3 - 4 386 58
AL834212.1 - 15 - 16 2098 58
AY074490.1 - 16 - 17 3420 58
AY074491.1 - 16 - 17 3484 58
BC007520.1 - 3 - 4 291 58

View Full Product Details