The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136205.2 | NP_001129677.1 | 4 - 5 | 1176 | 83 |
NM_001142730.2 | NP_001136202.1 | 4 - 5 | 2440 | 83 | |
NM_001258221.1 | NP_001245150.1 | 4 - 5 | 717 | 83 | |
NM_001258222.1 | NP_001245151.1 | 4 - 5 | 673 | 83 | |
NM_198991.3 | NP_945342.1 | 5 - 6 | 756 | 83 | |
XM_017025708.1 | XP_016881197.1 | 4 - 5 | 966 | 83 | |
GenBank mRNA | AF542549.1 | - | 5 - 6 | 763 | 83 |
AK289870.1 | - | 4 - 5 | 862 | 83 | |
BC063652.1 | - | 4 - 5 | 897 | 83 |