Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
Chromosome Location
Chr.17: 74274215 - 74314884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172810.1 NP_001166281.1 1 - 2 99 64
GenBank mRNA BC039582.1 - 1 - 2 89 64

View Full Product Details