Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
Chromosome Location
Chr.2: 159315312 - 159712439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289975.1 NP_001276904.1 15 - 16 3123 110
NM_013450.3 NP_038478.2 16 - 17 3231 110
XM_005246488.2 XP_005246545.2 16 - 17 3074 110
XM_005246489.4 XP_005246546.2 15 - 16 4177 110
XM_005246492.4 XP_005246549.2 15 - 16 4172 110
XM_006712461.3 XP_006712524.2 19 - 20 3648 110
XM_011511038.2 XP_011509340.1 16 - 17 3178 110
XM_011511039.1 XP_011509341.1 14 - 15 2828 110
XM_011511042.1 XP_011509344.1 14 - 15 2828 110
XM_011511043.1 XP_011509345.1 13 - 14 2726 110
XM_011511044.1 XP_011509346.1 13 - 14 2726 110
XM_011511045.1 XP_011509347.1 13 - 14 2633 110
XM_011511047.1 XP_011509349.1 15 - 16 2980 110
XM_011511048.2 XP_011509350.1 15 - 16 3045 110
XM_011511050.1 XP_011509352.1 13 - 14 2528 110
XM_011511051.1 XP_011509353.1 13 - 14 2720 110
XM_011511052.1 XP_011509354.1 12 - 13 2432 110
XM_011511053.2 XP_011509355.1 14 - 15 2828 110
XM_011511054.2 XP_011509356.1 14 - 15 2828 110
XM_011511056.1 XP_011509358.1 4 - 5 448 110
XM_017003918.1 XP_016859407.1 19 - 20 3638 110
XM_017003919.1 XP_016859408.1 19 - 20 3632 110
XM_017003920.1 XP_016859409.1 21 - 22 3849 110
XM_017003921.1 XP_016859410.1 16 - 17 3172 110
XM_017003922.1 XP_016859411.1 16 - 17 3068 110
XM_017003923.1 XP_016859412.1 14 - 15 2828 110
XM_017003924.1 XP_016859413.1 13 - 14 2720 110
XM_017003925.1 XP_016859414.1 15 - 16 3076 110
XM_017003926.1 XP_016859415.1 15 - 16 2940 110
XM_017003927.1 XP_016859416.1 15 - 16 2980 110
XM_017003928.1 XP_016859417.1 15 - 16 2884 110
XM_017003929.1 XP_016859418.1 15 - 16 2972 110
XM_017003930.1 XP_016859419.1 15 - 16 2780 110
XM_017003931.1 XP_016859420.1 15 - 16 2878 110
XM_017003932.1 XP_016859421.1 12 - 13 2531 110
XM_017003933.1 XP_016859422.1 14 - 15 2751 110
XM_017003934.1 XP_016859423.1 14 - 15 2646 110
XM_017003935.1 XP_016859424.1 13 - 14 2584 110
XM_017003936.1 XP_016859425.1 13 - 14 2544 110
XM_017003937.1 XP_016859426.1 8 - 9 1245 110
GenBank mRNA AB032255.1 - 11 - 12 2604 110
AB040909.2 - 15 - 16 3091 110
AK027612.1 - 7 - 8 858 110
AK127551.1 - 15 - 16 2641 110
AK307683.1 - 14 - 15 2468 110
AL834381.1 - 16 - 17 3126 110
BC012576.1 - 12 - 13 2243 110

View Full Product Details